This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hg05077929_m1
See other ALG9 GE Assays ›
Gene Symbol:
Alg9
Gene Name:
ALG9, alpha-1,2-mannosyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624784: 6936430 - 7049891 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID
Hg05077929_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Alg9
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004856537.2
XP_004856594.1
10 - 11
1284
64
XM_004856538.2
XP_004856595.1
9 - 10
1149
64
XM_004856539.3
XP_004856596.1
10 - 11
1284
64
XM_004856540.3
XP_004856597.1
10 - 11
1284
64
XM_004856542.3
XP_004856599.1
10 - 11
1284
64
XM_004856543.3
XP_004856600.1
10 - 11
1284
64
XM_004856544.2
XP_004856601.1
10 - 11
1284
64
XM_013076580.2
XP_012932034.1
10 - 11
1284
64
XM_021237887.1
XP_021093546.1
10 - 11
1439
64
XM_021237888.1
XP_021093547.1
10 - 11
1284
64
XM_021237889.1
XP_021093548.1
10 - 11
1284
64
XM_021237890.1
XP_021093549.1
10 - 11
1284
64
XM_021237891.1
XP_021093550.1
10 - 11
1284
64
XM_021237892.1
XP_021093551.1
10 - 11
1284
64
XM_021237893.1
XP_021093552.1
10 - 11
1284
64
XM_021237894.1
XP_021093553.1
10 - 11
1284
64
XM_021237895.1
XP_021093554.1
10 - 11
1284
64
XM_021237896.1
XP_021093555.1
10 - 11
1284
64
XM_021237897.1
XP_021093556.1
9 - 10
1173
64
XM_021237898.1
XP_021093557.1
9 - 10
1135
64
XM_021237899.1
XP_021093558.1
9 - 10
1368
64
XM_021237900.1
XP_021093559.1
7 - 8
954
64
XM_021237901.1
XP_021093560.1
10 - 11
1285
64
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Probe spans exons
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0