Assay Details
Gene Symbol: Dda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873234.3 | XP_004873291.1 | 3 - 4 | 279 | 64 | |
XM_021251122.1 | XP_021106781.1 | 4 - 5 | 294 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
