Assay Details
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864590.3 | XP_004864647.1 | 5 - 6 | 718 | 67 | |
XM_004864591.3 | XP_004864648.1 | 5 - 6 | 715 | 67 | ||
XM_021244238.1 | XP_021099897.1 | 5 - 6 | 489 | 67 | ||
XM_021244239.1 | XP_021099898.1 | 5 - 6 | 486 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
