This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05069244_m1
See other MRPL23 GE Assays ›
Gene Symbol:
Mrpl23
Gene Name:
mitochondrial ribosomal protein L23
Gene Aliases:
GW7_00392
Chromosome Location:
Chr.Un|NW_004624767: 14818175 - 14826620 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05069244_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004852090.3 XP_004852147.1 2 - 3 190 84
XM_004852091.3 XP_004852148.1 2 - 3 225 84
XM_013075007.2 XP_012930461.1 4 - 5 307 84
XM_021264412.1 XP_021120071.1 2 - 3 570 84
XM_021264413.1 XP_021120072.1 2 - 3 578 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0