Assay Details
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310262.1 | NP_001297191.1 | 4 - 5 | 566 | 64 | |
XM_004861144.3 | XP_004861201.1 | 5 - 6 | 852 | 64 | ||
XM_004861145.3 | XP_004861202.1 | 5 - 6 | 653 | 64 | ||
XM_004861146.2 | XP_004861203.1 | 4 - 5 | 693 | 64 | ||
XM_004861147.3 | XP_004861204.1 | 4 - 5 | 787 | 64 | ||
XM_004861148.2 | XP_004861205.1 | 4 - 5 | 732 | 64 | ||
XM_004861152.2 | XP_004861209.1 | 4 - 5 | 723 | 64 | ||
XM_004861153.3 | XP_004861210.1 | 3 - 4 | 554 | 64 | ||
XM_004861154.2 | XP_004861211.1 | 3 - 4 | 604 | 64 | ||
XM_013078291.2 | XP_012933745.1 | 5 - 6 | 682 | 64 | ||
XM_013078292.2 | XP_012933746.1 | 5 - 6 | 717 | 64 | ||
XM_013078293.2 | XP_012933747.1 | 4 - 5 | 554 | 64 | ||
XM_021241340.1 | XP_021096999.1 | 6 - 7 | 986 | 64 | ||
XM_021241341.1 | XP_021097000.1 | 6 - 7 | 888 | 64 | ||
XM_021241343.1 | XP_021097002.1 | 5 - 6 | 760 | 64 | ||
XM_021241344.1 | XP_021097003.1 | 4 - 5 | 590 | 64 | ||
GenBank mRNA | KM486777.1 | - | 4 - 5 | 566 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
