This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg01031061_m1
See other MRAS GE Assays ›
Gene Symbol:
Mras
Gene Name:
muscle RAS oncogene homolog
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624730: 14838847 - 14938682 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg01031061_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mras
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004834312.3 XP_004834369.1 5 - 6 801 80
XM_004834313.3 XP_004834370.1 5 - 6 650 80
XM_004834314.3 XP_004834371.1 5 - 6 991 80
XM_004834315.2 XP_004834372.1 5 - 6 599 80
XM_013070854.2 XP_012926308.1 5 - 6 654 80
XM_021248775.1 XP_021104434.1 4 - 5 354 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0