This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm07348739_s1
See other NLP4 GE Assays ›
Gene Symbol:
NLP4
Gene Name:
protein CUP-SHAPED COTYLEDON 2-like
Gene Aliases:
Glyma12g35530
Chromosome Location:
Chr.12: 38655462 - 38657254 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm07348739_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NLP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287453.1 NP_001274382.1 3 519 77
GenBank mRNA JX392401.1 - 3 519 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0