This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm07348665_s1
See other LOC100783345 GE Assays ›
Gene Symbol:
LOC100783345
Gene Name:
agamous-like MADS-box protein AGL12-like
Gene Aliases:
-
Chromosome Location:
Chr.12: 384333 - 389437 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm07348665_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100783345
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001255908.1 NP_001242837.1 1 113 130
GenBank mRNA BT099142.1 - 1 113 130

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BT099142.1, NM_001255908.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Glycine max Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0