Assay Details
Gene Symbol: LOC100792205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289303.1 | NP_001276232.1 | 3 - 4 | 486 | 78 | |
GenBank mRNA | AK244417.1 | - | 3 - 4 | 487 | 78 | |
AK286980.1 | - | 3 - 4 | 485 | 78 | ||
BT097954.1 | - | 3 - 4 | 488 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
