This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm07331473_mH
See other ICEE GE Assays ›
Gene Symbol:
ICEE
Gene Name:
transcription factor ICE1-like
Gene Aliases:
-
Chromosome Location:
Chr.16: 30422468 - 30426572 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm07331473_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ICEE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001254406.1 NP_001241335.1 3 - 4 1148 93
GenBank mRNA HM989929.1 - 3 - 4 1148 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Glycine max Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0