This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm07331087_g1
See other ANN14 GE Assays ›
Gene Symbol:
ANN14
Gene Name:
annexin A7-like
Gene Aliases:
-
Chromosome Location:
Chr.13: 29263571 - 29265111 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm07331087_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ANN14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001254318.1 NP_001241247.1 3 - 4 847 157
GenBank mRNA HM012593.1 - 3 - 4 847 157

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons

Panther Classification:

Molecular Function -

calcium-binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0