This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm03759609_m1
See other LOC732620 GE Assays ›
Gene Symbol:
LOC732620
Gene Name:
SNI1
Gene Aliases:
-
Chromosome Location:
Chr.13: 27442273 - 27447096 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm03759609_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC732620
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001250989.1 NP_001237918.1 13 - 14 1205 91
GenBank mRNA DQ468343.1 - 13 - 14 1205 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Glycine max Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0