This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm03758790_g1
See other HEMB GE Assays ›
Gene Symbol:
HEMB
Gene Name:
aminolevulinate, delta-, dehydratase
Gene Aliases:
ALADH, Alad
Chromosome Location:
Chr.4: 48357705 - 48363758 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm03758790_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HEMB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001251808.1 NP_001238737.1 8 - 9 857 95
GenBank mRNA AK243972.1 - 8 - 9 866 95
AK285742.1 - 7 - 8 1291 95
BT093896.1 - 1 - 2 268 95
U04525.1 - 8 - 9 857 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons

Panther Classification:

Molecular Function -

dehydratase lyase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0