This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gm03757425_g1
See other PCS1 GE Assays ›
Gene Symbol:
PCS1
Gene Name:
homo-phytochelatin synthase
Gene Aliases:
GmhPCS1
Chromosome Location:
Chr.3: 40002472 - 40005896 on Build Glycine_max_v2.1
Species:
Glycine max
Species Specific ID (Flybase ID):
-
Assay ID Gm03757425_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001248647.1 NP_001235576.1 6 - 7 1064 81
GenBank mRNA AF411075.1 - 6 - 7 1064 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0