Assay Details
Gene Symbol: LOC107049647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145913.1 | XP_025001681.1 | 2 - 3 | 235 | 75 | |
XM_025145914.1 | XP_025001682.1 | 2 - 3 | 1306 | 75 | ||
XM_025145915.1 | XP_025001683.1 | 2 - 3 | 209 | 75 | ||
XM_025145916.1 | XP_025001684.1 | 2 - 3 | 209 | 75 | ||
XM_025145917.1 | XP_025001685.1 | 1 - 2 | 1684 | 75 | ||
XM_025145918.1 | XP_025001686.1 | 1 - 2 | 1684 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
