This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07204364_m1
See other METTL21EP GE Assays ›
Gene Symbol:
METTL21EP
Gene Name:
methyltransferase like 21E, pseudogene
Gene Aliases:
METTL21CP1
Chromosome Location:
Chr.1: 144171312 - 144191502 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07204364_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL21EP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001232406.4 XP_001232407.2 2 - 3 279 149
XM_004938556.3 XP_004938613.1 3 - 4 357 149
XM_015275041.2 XP_015130527.1 3 - 4 357 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header