This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07197141_g1
See other COL2A1L GE Assays ›
Gene Symbol:
COL2A1L
Gene Name:
collagen alpha-1(II) chain-like
Gene Aliases:
-
Chromosome Location:
Chr.17: 2732326 - 2775803 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07197141_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COL2A1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015279683.2 XP_015135169.1 62 - 63 4567 158
XM_025156090.1 XP_025011858.1 62 - 63 4567 158
XM_025156091.1 XP_025011859.1 62 - 63 4564 158
XM_025156092.1 XP_025011860.1 61 - 62 4513 158
XM_025156093.1 XP_025011861.1 61 - 62 4513 158
XM_025156094.1 XP_025011862.1 60 - 61 4402 158
XM_025156095.1 XP_025011863.1 62 - 63 4567 158
XM_025156096.1 XP_025011864.1 62 - 63 4773 158
XM_025156097.1 XP_025011865.1 58 - 59 4424 158

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix protein extracellular matrix structural protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0