This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07188079_m1
See other TMEM200B GE Assays ›
Gene Symbol:
TMEM200B
Gene Name:
transmembrane protein 200B
Gene Aliases:
-
Chromosome Location:
Chr.23: 2817710 - 2841391 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07188079_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM200B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004947729.3 XP_004947786.1 1 - 2 1041 95
XM_025143131.1 XP_024998899.1 2 - 3 627 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004947729.3, XM_025143131.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0