This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07185192_m1
See other PLPP1 GE Assays ›
Gene Symbol:
PLPP1
Gene Name:
phospholipid phosphatase 1
Gene Aliases:
PPAP2A
Chromosome Location:
Chr.Z: 16804843 - 16861139 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07185192_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLPP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004937240.3 XP_004937297.1 5 - 6 906 105
XM_424730.6 XP_424730.3 5 - 6 875 105
GenBank mRNA BX934014.2 - 4 - 5 794 105
CR382434.1 - 6 - 7 895 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0