This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07183367_g1
See other ALMS1 GE Assays ›
Gene Symbol:
ALMS1
Gene Name:
ALMS1, centrosome and basal body associated protein
Gene Aliases:
-
Chromosome Location:
Chr.4: 89596437 - 89649576 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07183367_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALMS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015286051.2 XP_015141537.1 16 - 17 8536 103
XM_015286052.2 XP_015141538.1 16 - 17 8536 103
XM_015286053.2 XP_015141539.1 16 - 17 8464 103
XM_015286054.2 XP_015141540.1 16 - 17 7834 103
XM_015286055.2 XP_015141541.1 8 - 9 2265 103
XM_015286056.2 XP_015141542.1 8 - 9 2265 103
XM_025149803.1 XP_025005571.1 16 - 17 7831 103
XM_420887.6 XP_420887.5 16 - 17 8539 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm