Assay Details
Gene Symbol: CRELD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293349.2 | XP_015148835.2 | 6 - 7 | 952 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
