This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07182273_s1
See other DLGAP1L GE Assays ›
Gene Symbol:
DLGAP1L
Gene Name:
discs, large (Drosophila) homolog-associated protein 1-like
Gene Aliases:
-
Chromosome Location:
Chr.2: 100411775 - 100803477 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07182273_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLGAP1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015282556.2 XP_015138042.2 13 4387 101
XM_015282557.2 XP_015138043.2 14 4487 101
XM_015282558.2 XP_015138044.2 13 4189 101
XM_015282560.2 XP_015138046.2 12 4237 101
XM_025147883.1 XP_025003651.1 10 2942 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0