This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07175048_g1
See other TIMM23B GE Assays ›
Gene Symbol:
TIMM23B
Gene Name:
translocase of inner mitochondrial membrane 23 homolog B
Gene Aliases:
TIMM23
Chromosome Location:
Chr.6: 3488788 - 3507996 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07175048_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIMM23B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_421501.6 XP_421501.2 1 - 2 147 141
GenBank mRNA BX930913.1 - 1 - 2 131 141
BX933189.1 - 1 - 2 138 141
BX933971.2 - 1 - 2 108 141
BX935539.2 - 1 - 2 153 141
BX950355.2 - 1 - 2 153 141

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

amino acid transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0