This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07174855_g1
See other UBE2T GE Assays ›
Gene Symbol:
UBE2T
Gene Name:
ubiquitin conjugating enzyme E2 T
Gene Aliases:
-
Chromosome Location:
Chr.26: 1099102 - 1102466 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07174855_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2T
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003642716.4 XP_003642764.1 169 108
XM_003642717.4 XP_003642765.1 187 108
XM_003642718.4 XP_003642766.1 170 108
XM_003642719.4 XP_003642767.1 174 108
XM_015298812.2 XP_015154298.1 161 108
XM_015298813.2 XP_015154299.1 256 108
XM_025143845.1 XP_024999613.1 246 108
XM_025143846.1 XP_024999614.1 219 108
XM_419230.6 XP_419230.2 193 108
GenBank mRNA BX930586.1 - 146 108
BX930853.1 - 225 108
BX931719.2 - 156 108
BX932295.1 - 193 108
BX933693.1 - 159 108
BX950762.1 - 171 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0