This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07170851_s1
See other FRS3 GE Assays ›
Gene Symbol:
FRS3
Gene Name:
fibroblast growth factor receptor substrate 3
Gene Aliases:
-
Chromosome Location:
Chr.26: 4952169 - 4965491 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07170851_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FRS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004948431.3 XP_004948488.1 7125 84
XM_015298974.2 XP_015154460.1 7092 84
XM_015298976.2 XP_015154462.1 6951 84
XM_015298977.2 XP_015154463.1 6137 84
XM_015298978.2 XP_015154464.1 6246 84
XM_015298979.2 XP_015154465.1 6133 84
XM_015298980.2 XP_015154466.1 6125 84
XM_015298981.2 XP_015154467.1 5823 84
XM_015298982.2 XP_015154468.1 5811 84
XM_025143791.1 XP_024999559.1 7001 84
XM_025143792.1 XP_024999560.1 6242 84
GenBank mRNA CR523802.1 - 414 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0