This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07168276_m1
See other CABP1 GE Assays ›
Gene Symbol:
CABP1
Gene Name:
calcium binding protein 1
Gene Aliases:
CABP5
Chromosome Location:
Chr.15: 9491805 - 9541203 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07168276_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CABP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001233508.5 XP_001233509.2 6 - 7 1002 65
XM_004934386.3 XP_004934443.2 5 - 6 827 65
XM_004934387.3 XP_004934444.1 4 - 5 967 65
XM_015275412.2 XP_015130898.1 5 - 6 1293 65
XM_015275413.2 XP_015130899.1 4 - 5 1127 65
XM_025155731.1 XP_025011499.1 5 - 6 710 65
XM_025155732.1 XP_025011500.1 4 - 5 536 65
XM_025155733.1 XP_025011501.1 4 - 5 553 65
GenBank mRNA CR391151.1 - 4 - 5 573 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0