Assay Details
| Gene Symbol: DNAH5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282404.2 | XP_015137890.2 | 74 - 75 | 13542 | 111 | |
| GenBank mRNA | CR387609.1 | - | 4 - 5 | 800 | 111 | |
More Information
Set Membership: |
|

DNAH5L
Genomic Map