Assay Details
Gene Symbol: DNAH5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282404.2 | XP_015137890.2 | 73 - 74 | 13335 | 109 | |
GenBank mRNA | CR387609.1 | - | 3 - 4 | 593 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
