This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07163247_s1
See other WDR89 GE Assays ›
Gene Symbol:
WDR89
Gene Name:
WD repeat domain 89
Gene Aliases:
-
Chromosome Location:
Chr.5: 53432489 - 53482285 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07163247_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WDR89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277546.3 NP_001264475.1 2 1021 90
XM_015287497.2 XP_015142983.1 3 1175 90
XM_015287498.2 XP_015142984.1 2 1850 90
XM_015287499.2 XP_015142985.1 2 1162 90
XM_025150799.1 XP_025006567.1 4 2145 90
XM_025150800.1 XP_025006568.1 4 2145 90
XM_025150801.1 XP_025006569.1 4 2145 90
XM_025150802.1 XP_025006570.1 2 1809 90
XM_025150803.1 XP_025006571.1 3 4738 90
XM_025150804.1 XP_025006572.1 4 1887 90
XM_025150805.1 XP_025006573.1 2 1029 90
XM_025150806.1 XP_025006574.1 3 1249 90
XM_025150807.1 XP_025006575.1 4 1285 90
XM_025150808.1 XP_025006576.1 3 1209 90
XM_025150809.1 XP_025006577.1 2 4620 90
GenBank mRNA BX933064.2 - 2 1021 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0