This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07162278_g1
See other S100A12 GE Assays ›
Gene Symbol:
S100A12
Gene Name:
S100 calcium binding protein A12
Gene Aliases:
S100A9
Chromosome Location:
Chr.25: 3296969 - 3298404 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07162278_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001305151.1 NP_001292080.1 1 - 2 50 104
GenBank mRNA X61200.1 - 55 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001305151.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

xenobiotic metabolic process
killing of cells of other organism
positive regulation of I-kappaB kinase/NF-kappaB signaling
defense response to fungus
antimicrobial humoral immune response mediated by antimicrobial peptide
calcium ion binding
calcium-dependent protein binding
RAGE receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0