This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07161331_gH
See other IGLL1 GE Assays ›
Gene Symbol:
IGLL1
Gene Name:
immunoglobulin lambda-like polypeptide 1
Gene Aliases:
IGLV, IgL
Chromosome Location:
Chr.15: 8213817 - 8231115 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07161331_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGLL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278545.1 NP_001265474.1 1 - 2 89 106
XM_015275314.2 XP_015130800.1 1 - 2 84 106
XM_015275315.2 XP_015130801.1 1 - 2 84 106
XM_015275316.2 XP_015130802.1 1 - 2 84 106
XM_015275317.2 XP_015130803.1 1 - 2 84 106
XM_025155490.1 XP_025011258.1 1 - 2 84 106
XM_025155491.1 XP_025011259.1 1 - 2 84 106
XM_025155492.1 XP_025011260.1 1 - 2 84 106
XM_025155493.1 XP_025011261.1 1 - 2 84 106
XM_025155494.1 XP_025011262.1 1 - 2 84 106
XM_025155495.1 XP_025011263.1 1 - 2 84 106
XM_025155496.1 XP_025011264.1 1 - 2 84 106
XM_025155498.1 XP_025011266.1 1 - 2 84 106
GenBank mRNA BX929742.1 - 1 - 2 74 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001278545.1, XM_015275314.2, XM_015275315.2, XM_015275316.2, XM_015275317.2, XM_025155490.1, XM_025155491.1, XM_025155492.1, XM_025155493.1, XM_025155494.1, XM_025155495.1, XM_025155496.1, XM_025155498.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s)

immunoglobulin production
immune response
complement activation
complement activation, classical pathway

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0