Assay Details
| Gene Symbol: SDHAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277889.1 | NP_001264818.1 | 3 - 4 | 399 | 100 | |
| NM_001277890.1 | NP_001264819.1 | 3 - 4 | 375 | 100 | ||
| XM_015286085.2 | XP_015141571.2 | 2 - 3 | 512 | 100 | ||
| GenBank mRNA | BU205400.1 | - | 3 - 4 | 378 | 100 | |
| BU353870.1 | - | 3 - 4 | 398 | 100 | ||
| CN228764.1 | - | 3 - 4 | 354 | 100 | ||
More Information
Set Membership: |
|

SDHAF2
Genomic Map