This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07160847_m1
See other PERPB GE Assays ›
Gene Symbol:
PERPB
Gene Name:
p53 apoptosis effector related to PMP22 B
Gene Aliases:
PERP, PERP2
Chromosome Location:
Chr.3: 54597280 - 54606430 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07160847_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PERPB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277704.1 NP_001264633.1 1 - 2 308 89
GenBank mRNA BX930445.2 - 1 - 2 268 89
BX930611.2 - 1 - 2 276 89
BX933259.1 - 1 - 2 256 89
DN850405.1 - 1 - 2 308 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0