This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg07158169_g1
See other CCL4 GE Assays ›
Gene Symbol:
CCL4
Gene Name:
chemokine (C-C motif) ligand 4
Gene Aliases:
MIP-1beta, SCYA4
Chromosome Location:
Chr.19: 569498 - 570362 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg07158169_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030360.2 NP_001025531.2 2 - 3 245 86
GenBank mRNA AJ243034.1 - 2 - 3 245 86
CR354001.1 - 2 - 3 86
L34553.1 - 2 - 3 152 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

monocyte chemotaxis
inflammatory response
G-protein coupled receptor signaling pathway
neutrophil chemotaxis
positive regulation of GTPase activity
eosinophil chemotaxis
lymphocyte chemotaxis
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
chemokine activity
CCR chemokine receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0