This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03644279_m1
See other TFAP2D GE Assays ›
Gene Symbol:
TFAP2D
Gene Name:
transcription factor AP-2 delta
Gene Aliases:
-
Chromosome Location:
Chr.3: 108262723 - 108317366 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03644279_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TFAP2D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135785.1 NP_001129257.1 4 - 5 763 66
XM_015285152.2 XP_015140638.1 3 - 4 1766 66
XM_015285153.2 XP_015140639.1 4 - 5 1606 66
XM_015285154.2 XP_015140640.1 4 - 5 1383 66
GenBank mRNA EU740390.1 - 4 - 5 763 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0