This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03370418_g1
See other SDC4 GE Assays ›
Gene Symbol:
SDC4
Gene Name:
syndecan 4
Gene Aliases:
syndecan-4
Chromosome Location:
Chr.20: 5127360 - 5141905 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03370418_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007869.1 NP_001007870.1 3 - 4 252 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
regulation of fibroblast migration
cell migration
negative regulation of T cell proliferation
wound healing, spreading of cells
inner ear receptor stereocilium organization
positive regulation of protein tyrosine kinase activity
activation of GTPase activity
negative regulation of skeletal muscle satellite cell proliferation
negative regulation of satellite cell differentiation
skeletal muscle satellite cell migration
regulation of protein localization to plasma membrane
positive regulation of protein localization to plasma membrane
positive regulation of exosomal secretion
positive regulation of extracellular exosome assembly
identical protein binding
thrombospondin receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0