This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03368488_m1
See other ALDH3A2 GE Assays ›
Gene Symbol:
ALDH3A2
Gene Name:
aldehyde dehydrogenase 3 family member A2
Gene Aliases:
-
Chromosome Location:
Chr.19: 6988772 - 7000330 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03368488_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH3A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006223.1 NP_001006223.1 3 - 4 651 83
XM_015295773.2 XP_015151259.2 5 - 6 5619 83
XM_015295774.2 XP_015151260.2 4 - 5 647 83
GenBank mRNA AJ719792.1 - 3 - 4 651 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0