Assay Details
Gene Symbol: SH3GLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006200.1 | NP_001006200.1 | 1 - 2 | 126 | 63 | |
XM_015279476.2 | XP_015134962.1 | 1 - 2 | 359 | 63 | ||
XM_015279478.2 | XP_015134964.1 | 1 - 2 | 107 | 63 | ||
XM_015279479.2 | XP_015134965.1 | 1 - 2 | 353 | 63 | ||
XM_015279480.2 | XP_015134966.1 | 1 - 2 | 107 | 63 | ||
XM_015279481.2 | XP_015134967.1 | 1 - 2 | 356 | 63 | ||
XM_015279482.2 | XP_015134968.1 | 1 - 2 | 160 | 63 | ||
XM_025156074.1 | XP_025011842.1 | 1 - 2 | 110 | 63 | ||
GenBank mRNA | AJ720553.1 | - | 1 - 2 | 126 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
