This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03366722_m1
See other CTNNA2 GE Assays ›
Gene Symbol:
CTNNA2
Gene Name:
catenin alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 87426722 - 87880923 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03366722_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTNNA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205136.1 NP_990467.1 8 - 9 1265 82
XM_015285873.2 XP_015141359.1 8 - 9 1186 82
XM_025149520.1 XP_025005288.1 7 - 8 1144 82
XM_025149521.1 XP_025005289.1 2 - 3 878 82
XM_025149522.1 XP_025005290.1 2 - 3 195 82
XM_025149523.1 XP_025005291.1 7 - 8 1143 82
XM_025149524.1 XP_025005292.1 7 - 8 1143 82
GenBank mRNA D11090.1 - 8 - 9 1265 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
axonogenesis
protein localization
epithelial cell differentiation
dendrite morphogenesis
brain morphogenesis
regulation of synapse structural plasticity
trigeminal ganglion formation
cell-cell adhesion
structural molecule activity
protein binding
cadherin binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0