This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03364850_m1
See other LSP1P1 GE Assays ›
Gene Symbol:
LSP1P1
Gene Name:
lymphocyte-specific protein 1 pseudogene 1
Gene Aliases:
LSP1
Chromosome Location:
Chr.5: 14250723 - 14289845 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03364850_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LSP1P1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303036.1 NP_001289965.1 10 - 11 1182 68
XM_015286521.1 XP_015142007.1 10 - 11 1419 68
XM_025150541.1 XP_025006309.1 11 - 12 1299 68
GenBank mRNA AB101641.1 - 9 - 10 1003 68
AJ851520.1 - 10 - 11 1182 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0