This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03364078_m1
See other KHDRBS1 GE Assays ›
Gene Symbol:
KHDRBS1
Gene Name:
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases:
-
Chromosome Location:
Chr.23: 5230212 - 5251255 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03364078_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KHDRBS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204230.1 NP_989561.1 6 - 7 1173 65
XM_015297597.2 XP_015153083.1 6 - 7 1396 65
GenBank mRNA AY057837.1 - 6 - 7 1173 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
RNA splicing
negative regulation of transcription, DNA-templated
positive regulation of translational initiation
regulation of RNA export from nucleus
positive regulation of RNA export from nucleus
T cell receptor signaling pathway
RNA binding
mRNA binding
poly(A) binding
SH3 domain binding
protein tyrosine kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0