This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03363286_m1
See other EDNRB2 GE Assays ›
Gene Symbol:
EDNRB2
Gene Name:
endothelin receptor B subtype 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 11158651 - 11169803 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03363286_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EDNRB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_204120.1 NP_989451.1 5 - 6 1043 65
XM_015278144.2 XP_015133630.1 4 - 5 986 65
XM_015278145.2 XP_015133631.1 4 - 5 986 65
XM_015278146.2 XP_015133632.1 4 - 5 1041 65
XM_015278148.2 XP_015133634.1 5 - 6 1062 65
XM_015278149.2 XP_015133635.1 5 - 6 1036 65
XM_015278150.2 XP_015133636.1 4 - 5 978 65
XM_015278151.2 XP_015133637.1 5 - 6 1145 65
GenBank mRNA AB697059.1 - 3 - 4 766 65
AB697062.1 - 3 - 4 766 65
AF472617.1 - 5 - 6 1043 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0