This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03362131_m1
See other MSANTD2 GE Assays ›
Gene Symbol:
MSANTD2
Gene Name:
Myb/SANT DNA binding domain containing 2
Gene Aliases:
C11ORF61, C24H11orf61
Chromosome Location:
Chr.24: 297221 - 317771 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03362131_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109786.1 NP_001103256.1 2 - 3 772 123
XM_015298049.2 XP_015153535.1 3 - 4 843 123
XM_015298050.2 XP_015153536.1 3 - 4 840 123
XM_015298051.1 XP_015153537.1 2 - 3 5825 123
XM_015298053.1 XP_015153539.1 3 - 4 5870 123
XM_015298054.1 XP_015153540.1 3 - 4 5867 123
XM_025143212.1 XP_024998980.1 3 - 4 5750 123
GenBank mRNA AJ721009.1 - 2 - 3 772 123

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0