This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03361495_m1
See other ARPP19 GE Assays ›
Gene Symbol:
ARPP19
Gene Name:
cAMP regulated phosphoprotein 19
Gene Aliases:
ARPP-19
Chromosome Location:
Chr.10: 9178651 - 9190069 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03361495_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARPP19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081707.1 NP_001075176.1 2 - 3 353 150
XM_025153873.1 XP_025009641.1 2 - 3 372 150
GenBank mRNA AJ719596.1 - 2 - 3 353 150

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
mitotic cell cycle
negative regulation of phosphoprotein phosphatase activity
negative regulation of protein dephosphorylation
positive regulation of gluconeogenesis
cell division
protein phosphatase inhibitor activity
receptor binding
potassium channel regulator activity
phosphatase inhibitor activity
protein phosphatase regulator activity
protein phosphatase 2A binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0