This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03360950_m1
See other CHAF1A GE Assays ›
Gene Symbol:
CHAF1A
Gene Name:
chromatin assembly factor 1 subunit A
Gene Aliases:
CAF-1P150, CAF-I-p150, CAF-Ip150
Chromosome Location:
Chr.28: 2605893 - 2620742 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03360950_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHAF1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079752.2 NP_001073220.2 14 - 15 2863 97
XM_015299949.2 XP_015155435.1 14 - 15 2847 97
GenBank mRNA AB195692.1 - 13 - 14 2717 97
AJ442963.1 - 2 - 3 149 97
AJ720199.1 - 14 - 15 2774 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s)

DNA replication
DNA repair
nucleosome assembly
cell cycle
chromatin assembly

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0