Assay Details
Gene Symbol: LOC428335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039321.1 | NP_001034410.1 | 11 - 12 | 1451 | 81 | |
XM_015299944.2 | XP_015155430.1 | 11 - 12 | 81 | |||
GenBank mRNA | AJ720427.1 | - | 11 - 12 | 1451 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
