This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03358015_m1
See other ALG12 GE Assays ›
Gene Symbol:
ALG12
Gene Name:
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases:
6-mannosyltransferase), alpha-1
Chromosome Location:
Chr.1: 19029127 - 19044919 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03358015_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALG12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039270.2 NP_001034359.2 4 - 5 565 76
XM_015272479.2 XP_015127965.1 5 - 6 76
XM_015272538.2 XP_015128024.1 5 - 6 76
XM_015272591.2 XP_015128077.1 5 - 6 76
XM_025147486.1 XP_025003254.1 4 - 5 76
GenBank mRNA AJ448794.1 - 4 - 5 565 76
AJ719392.1 - 4 - 5 564 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0