This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03357380_m1
See other C4BPG GE Assays ›
Gene Symbol:
C4BPG
Gene Name:
complement component 4 binding protein, GPI-anchored
Gene Aliases:
CR1L, CREG
Chromosome Location:
Chr.26: 2658111 - 2663820 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03357380_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C4BPG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033643.1 NP_001028815.1 1446 103
GenBank mRNA AB109024.1 - 1446 103
BX950749.2 - 800 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0