This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03355379_g1
See other GPR89A GE Assays ›
Gene Symbol:
GPR89A
Gene Name:
G protein-coupled receptor 89A
Gene Aliases:
GPR89, GPR89B
Chromosome Location:
Chr.1: 93899941 - 93923231 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03355379_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR89A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030791.1 NP_001025962.1 13 - 14 1201 83
XM_025149429.1 XP_025005197.1 13 - 14 1413 83
XM_025149433.1 XP_025005201.1 13 - 14 1494 83
GenBank mRNA AJ851452.1 - 13 - 14 1201 83
CR523472.1 - 6 - 7 551 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein transport
inorganic anion transport
ion transmembrane transport
regulation of ion transmembrane transport
skin development
intracellular pH reduction
voltage-gated anion channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0