Assay Details
Gene Symbol: CD200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030787.1 | NP_001025958.1 | 4 - 5 | 778 | 109 | |
XM_025149353.1 | XP_025005121.1 | 4 - 5 | 921 | 109 | ||
GenBank mRNA | AJ720088.1 | - | 4 - 5 | 778 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
